Orders LinkContactUsLinkComponent
Extraction thimbles, Munktell
Extraction thimbles, Munktell
33×80 mm
Catalog # 537-4528
Supplier:   Ahlstrom
Extraction thimbles, Munktell
Catalog # 537-4528
Supplier:  Ahlstrom
Extraction thimbles, Munktell
Extraction thimbles, Munktell
33×80 mm
Price
109,00 €
Log in for availability
1 * 25 items
Product Details & Documents
Grade ET/401, cellulose.
Particulary suitable for the extraction of organic combinations out of reaction mixtures, food, natural substances, lacquer and binder analysis in paint colour, dust and tar determination in gases.
Max. Temperature: 120 °C
Specifications
  • Int.Ø×H:
    33×80 mm