ContactUsLinkComponent
Accessories for Multiple Use Earclips, KLEENGUARD* H50
Catalog # 111-0141
Supplier:  HONEYWELL SAFETY
undefined
Accessories for Multiple Use Earclips, KLEENGUARD* H50
Catalog # 111-0141
Supplier:  HONEYWELL SAFETY
Specifications
  • Colour:
    Blue/orange
  • Description:
    Earplugs, with cord
  • Size:
    U*
  • Environmentally Preferable:
  • Pk:
    100 PAIR
Specifications
About this item
These earplugs offer a high visibility and metal detection.
  • T-shape for easy handling and comfortable long-term use
  • Self-adjusting foam expands to fit virtually any wearer
  • Non ferrous metal and bright colours
SNR: 35 dB
Certifications: EN 352-2
Caution: * One size fits all.