Petri dish racks, Nalgene®
391-2161
- Description:Petri dish rack
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:NAR Certificate of Retirement
- Pk:1
Petri dish rack with PC frame and white PC posts.
- Smooth, rounded corners
- Finger-grip handle and rubber feet make carrying safe and secure
- Rack can be decontaminated by heat sterilisation
For up to 54 plastic round dishes (100×15 mm) or 42 glass round dishes (100×15 mm). Simple assembly required.
W×D×H: 203×343×235 mm