Orders LinkContactUsLinkComponent
Stopcocks for desiccators
Stopcocks for desiccators
Supplier:   Lenz Laborglas
Stopcocks for desiccators
Supplier:  Lenz Laborglas
Certificates
Stopcocks for desiccators
Stopcocks for desiccators
2 Options Available Below
Product Details & Documents
With PTFE spindles (except stopper).
Product Family Options
Product Information
  • For
    Availability
    Price
  • 467-1254
    Needle-valve stopcock for desiccators, for lid tubulation, NS24/29 drip tip cone
    Needle-valve stopcock for desiccators, for lid tubulation, NS24/29 drip tip cone
    NOVUS
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 52,10 €
    52,10 €
    1 * 1 items
       52,10 €
  • 467-1253
    Needle-valve stopcock for desiccators, for side tubulation, NS24/29 cone
    Needle-valve stopcock for desiccators, for side tubulation, NS24/29 cone
    NOVUS
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 43,40 €
    43,40 €
    1 * 1 items
       43,40 €