ContactUsLinkComponent
Cover glasses for hæmacytometers
Cover glasses for hæmacytometers
Catalog # 631-0393
Supplier:  MARIENFELD
undefined
Cover glasses for hæmacytometers
Catalog # 631-0393
Supplier:  MARIENFELD
Specifications
  • Description:
    Cover glasses for counting chambers and haemacytometers
  • L×W:
    32×32 mm
  • Material:
    Glass
  • Thickness:
    0,4 mm
  • Pk:
    10
Specifications
About this item
Cover glasses for counting chambers and haemacytometers suitable for IVD applications.
  • Made from special optical glass
  • Ready for use
  • Ground and polished edges
Certifications: IVD directive 98/79/EC.
Ordering information: Include a batch number for traceability.
Packaging: 10 pieces in cardboard box, 10 cardboard boxes in a carton.