Orders LinkContactUsLinkComponent
Spray bottle
Spray bottle
Sprayer for chromatography, with 100 ml Erlenmeyer flask, ST 19/26, with aspirator bulb
Catalog # 552-6001
Supplier:   witeg Labortechnik
Spray bottle
Catalog # 552-6001
Supplier:  witeg Labortechnik
Spray bottle
Spray bottle
Sprayer for chromatography, with 100 ml Erlenmeyer flask, ST 19/26, with aspirator bulb
Price
71,50 €
Log in for availability
1 * 1 items
Product Details & Documents
Glass spray bottle with rubber bulb for simple manual operation of the solution spray.
  • Connection for compressed air generator
100 ml capacity
Specifications
  • Description:
    Sprayer for chromatography, with 100 ml Erlenmeyer flask, ST 19/26, with aspirator bulb