Orders LinkContactUsLinkComponent
Raschig rings
Raschig rings
Supplier:   Lenz Laborglas
Raschig rings
Supplier:  Lenz Laborglas
Certificates
Raschig rings
6 Options Available Below
Product Details & Documents
Soda lime glass.
  • For packing columns
  • Good chemical resistance
  • Good thermal resistance
Product Family Options
Product Information
  • Ext.Ø
    Height
    Availability
    Price
  • LENZ05127003
    Raschig ring
    Raschig ring
    3 mm
    3 mm
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 190,00 €
    190,00 €
    1 * 1 kg
       190,00 €
  • LENZ05127004
    Raschig ring
    Raschig ring
    4 mm
    4 mm
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 141,00 €
    141,00 €
    1 * 1 kg
       141,00 €
  • LENZ05127005
    Raschig ring
    Raschig ring
    5 mm
    5 mm
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 126,00 €
    126,00 €
    1 * 1 kg
       126,00 €
  • LENZ05127006
    Raschig ring
    Raschig ring
    6 mm
    6 mm
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 79,60 €
    79,60 €
    1 * 1 kg
       79,60 €
  • LENZ05127008
    Raschig ring
    Raschig ring
    8 mm
    8 mm
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 78,70 €
    78,70 €
    1 * 1 kg
       78,70 €
  • 201-1164
    Raschig ring
    Raschig ring
    10 mm
    10 mm
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 76,20 €
    76,20 €
    1 * 1 kg
       76,20 €