Sleeve protectors, Secutex® pro, PP/PE
Catalog # ZVGA30241-90
Supplier: Zellstoff-Vertriebs-GmbH & Co. KG
Specifications
- Garment application:Laboratory
- Gender:
- Colour:White
- Length:380 mm
- Environmentally Preferable:
- Pk:100 PAIR
Specifications
About this item
These laminate sleeve protectors provide protection against fine dust and aggressive and toxic substances. Fitted with two elasticated cuffs.
- Lint-free, resistant to abrasion, tear and water
- Permeable to water vapour which ensures breathability
- Anti-static