Overalls, ProShield® 20, model CHF5
Supplier: DuPont
Certificates
About this item
These SMS garments are typically used, depending on toxicity and exposure conditions, for protection against particles (Type 5), limited liquid splashes or sprays (Type 6). They are an ideal choice for workers seeking protection against dirt and grime during light duty work.
- Nylon zipper with flap
- Elasticated 2-piece hood, waist, wrists, and ankles
- Stitched external seams
- Anti-static treatment (EN 1149-5) - on inside in blue and on both sides in white
Colour: white or blue
Weight: 43 g/m²
Density: 62 g/m²
Type 5/6, EN 1149, EN 1073-2; Cat. III
Certifications: (EU) 2016/425, Type 5/6, EN 1149-5, EN 1073-2; Cat. III.
Specifications
- Catalog No:
- 113-8367
- 113-8377
- 113-8378
- 113-8375
- 113-8374
- 113-8373
- 113-8376
- 113-8369
- 113-8372
- 113-8368
- 113-8371
- 113-8370
- Model:
- 20 - CHF5
- 20 - CHF5
- 20 - CHF5
- 20 - CHF5
- 20 - CHF5
- 20 - CHF5
- 20 - CHF5
- 20 - CHF5
- 20 - CHF5
- 20 - CHF5
- 20 - CHF5
- 20 - CHF5
- Anti-static:
- +
- +
- +
- +
- +
- +
- +
- +
- +
- +
- +
- +
- Garment application:
- General Purpose, Laboratory, Industrial
- General Purpose, Laboratory, Industrial
- General Purpose, Laboratory, Industrial
- General Purpose, Laboratory, Industrial
- General Purpose, Laboratory, Industrial
- General Purpose, Laboratory, Industrial
- General Purpose, Laboratory, Industrial
- General Purpose, Laboratory, Industrial
- General Purpose, Laboratory, Industrial
- General Purpose, Laboratory, Industrial
- General Purpose, Laboratory, Industrial
- General Purpose, Laboratory, Industrial
- Weight:
- 43 g/m²
- 43 g/m²
- 43 g/m²
- 43 g/m²
- 43 g/m²
- 43 g/m²
- 43 g/m²
- 43 g/m²
- 43 g/m²
- 43 g/m²
- 43 g/m²
- 43 g/m²