Aprons, Secutex® pro, Z 40, PP/PE
White
Catalog # 114-3441
Supplier: Zellstoff-Vertriebs-GmbH & Co. KG
Jump to:
Product Details & Documents
This disposable apron made from laminated anti-static PP/PE protects against aggressive and toxic substances.
- Lint-free, water and particle-proof
- Long version with neckband and belt
Colour: white
Specifications
- Colour:White
- Cat. No.:114-3441
- Garment application:Chemical protection