Orders LinkContactUsLinkComponent
Threaded closures for rectangular bottles
Threaded closures for rectangular bottles
Threaded closures for rectangular bottles
PP, white.
  • Tamper-evident closure
  • Self-sealing with internal cone
  • Can only be used for rectangular bottles 215-8331 to 215-8334
Product Family Options
  • Description
    Colour
  • Tamper-proof Screw-caps
    White
  • Screw cap, PP 28
    White
    1 * 20 items
    1 * 20 items - 14,60 €
    14,60 €
    1 * 20 items
       14,60 €