Orders LinkContactUsLinkComponent
Forceps, Nalgene®
Forceps, Nalgene®
Blue
Catalog # 229-0301
   Low Manufacturing Impact
Forceps, Nalgene®
Catalog # 229-0301
   Low Manufacturing Impact
Forceps, Nalgene®
Forceps, Nalgene®
Blue
Price
45,00 €
Log in for availability
1 * 12 items
Product Details & Documents
PP, blue. With double-action hinges for extra leverage. The jaws have interlocking teeth for holding objects securely. Scissor type handles with catch for locking the forceps. Can be used as clips for chromatography columns. Also serve as excellent tube clamps.
  • Good chemical resistance
  • Very good thermal resistance, autoclavable
  • Convenient and easy to use, durable
Specifications
  • Colour:
    Blue
  • Cat. No.:
    229-0301
  • Environmental certification:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Low Manufacturing Impact