Orders LinkContactUsLinkComponent
VWR®, Clamps
VWR®, Clamps
22 - 70 mm
Catalog # 241-0032
Supplier:   VWR Collection
VWR®, Clamps
Catalog # 241-0032
Supplier:  VWR Collection
VWR®, Clamps
VWR®, Clamps
22 - 70 mm
Price
69,80 €
Log in for availability
1 * 2 items
Product Details & Documents
Laboral aluminium alloy.
  • Round jaws covered in vinyl to protect clamped items
  • Two easy action knurled screws for adjusting each of the movable clamping jaws independently of each other
Specifications
  • Spanning width:
    22 - 70 mm
Accessories
Product Information
  • Catalog #
    For
    Availability
    Price
  • Protecting clamp for clamp jaws
    Protecting clamp for clamp jaws
    241-0149
    Clamps 241-0032
    Log in for availability
    1 * 10 items - 15,40 €