Orders LinkContactUsLinkComponent
Burette clamps
Burette clamps
Supplier:   BOCHEM
Burette clamps
Supplier:  BOCHEM
Certificates
Burette clamps
2 Options Available Below
Product Details & Documents
Stainless steel. For simple and secure fixing of burettes.
  • With bosshead for fixing to retort stand
  • Clamping range 0 to 20 mm
  • Jaws are covered with a silicone coating to protect the burettes
Product Family Options
Product Information
  • Spanning width
    For
    Availability
    Price
  • 0 - 20 mm
    One burette
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 54,00 €
    54,00 €
    1 * 1 items
       54,00 €
  • 0 - 20 mm
    Two burettes
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 74,00 €
    74,00 €
    1 * 1 items
       74,00 €