Orders LinkContactUsLinkComponent
Burettes, Nalgene®
Burettes, Nalgene®
Burettes, Nalgene®
Burettes, Nalgene®
Crystal-clear acrylic, PMP tip and TFE stopcock and plug.
  • Unaffected by dilute mineral acids and bases except HF and NH₄OH
  • Not designed for alcohol and organic solvents
Durable, excellent for general use, schools, industrial labs and field use.
 : ASTM E287
  • Catalog No:
  • 612-2558
  • 612-2559
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
Product Family Options
  • Capacity
    Division
    Tolerance
  • 25 ml
    0,1 ml
    0,06 ± ml
    1 * 1 items
    1 * 1 items - 175,00 €
    175,00 €
    1 * 1 items
       175,00 €
      Low Manufacturing Impact
    Specifications
    Cat. No.
    612-2558
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 50 ml
    0,1 ml
    0,1 ± ml
    1 * 1 items
    1 * 1 items - 202,00 €
    202,00 €
    1 * 1 items
       202,00 €
      Low Manufacturing Impact
    Specifications
    Cat. No.
    612-2559
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
Accessories
  •  
    Description
  • NALG71-3650-1025
    Stopcock assembly for burette, 25 ml
    1 * 2 items - 342,00 €