Slide tray
Catalog # 631-9002
Supplier: BERGMANN KARTON
Specifications
- No. of slides:20
- Colour:White
- Description:Slide tray
- Material:Cardboard
- Pk:1
Specifications
About this item
Made of strong cardboard, floorboard covered with white chrome paper, this board is designed for the storage of slides.