Orders LinkContactUsLinkComponent
Cryostorage boxes, arctic Square™
Cryostorage boxes, arctic Square™
Supplier:   HEATHROW
Cryostorage boxes, arctic Square™
Supplier:  HEATHROW
Certificates
Cryostorage boxes, arctic Square™
Cryostorage boxes, arctic Square™
3 Options Available Below
Product Details & Documents
PC, autoclavable.
  • The forward-sloped base and high-contrast, imprinted indexing on the transparent lid ensure quick visual orientation
  • All boxes include vent and drainage holes and are stackable
  • Alphanumeric identification on the lid
Safely store vials from −196 to +121 °C for repeated use in mechanical freezers as well as liquid nitrogen. Designed with unique features to help the user orient, identify, and access vials with ease. The lid is hinged to allow easy one-handed access to samples and a built-in stop prevents the lid from swinging too far when the box is picked up by the lid, helping to prevent accidental spills (5×5 array does not have hinged lid).
Documents
Product Family Options
Product Information
  • Compartments
    For
    Colour
    Well Ø
    W×D×H ext.
    Availability
    Price
  • 25 (5×5)
    Vial size up 2,0 ml
    Red
    12,5 mm
    76×76×53 mm
    Log in for availability
    Log in for availability
    1 * 8 items
    1 * 8 items - 98,80 €
    98,80 €
    1 * 8 items
       98,80 €
  • 81 (9×9)
    Vial size up 2,0 ml
    Assorted
    12,5 mm
    133×133×53 mm
    Log in for availability
    Log in for availability
    1 * 4 items
    1 * 4 items - 80,70 €
    80,70 €
    1 * 4 items
       80,70 €
  • 100 (10×10)
    Vial size up 2,0 ml
    Blue
    12,5 mm
    133×133×53 mm
    Log in for availability
    Log in for availability
    1 * 4 items
    1 * 4 items - 80,80 €
    80,80 €
    1 * 4 items
       80,80 €