Orders LinkContactUsLinkComponent
Screw caps for Jerricans
Screw caps for Jerricans
Supplier:   Kautex
Screw caps for Jerricans
Supplier:  Kautex
Certificates
Screw caps for Jerricans
Screw caps for Jerricans
12 Options Available Below
Product Details & Documents
Documents
Product Family Options
Screw closure HDPE, black, with tamper evident ring and venting system
Product Information
  • Thread
    For
    Availability
    Price
  • 45 mm
    2, 2,5, 3, 6 and 8 L jerricans
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 3,80 €
    3,80 €
    1 * 1 items
       3,80 €
  • 50 mm
    5 and 10 L jerricans
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 3,80 €
    3,80 €
    1 * 1 items
       3,80 €
  • 60 mm
    20/25 and 30 L jerricans
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 4,70 €
    4,70 €
    1 * 1 items
       4,70 €
Screw closure HDPE, black, with tamper evident ring
Product Information
  • Thread
    For
    Availability
    Price
  • 45 mm
    2/2,5/3/6 and 8 L jerricans
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 0,70 €
    0,70 €
    1 * 1 items
       0,70 €
  • 50 mm
    5 and 10 L jerricans
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 0,80 €
    0,80 €
    1 * 1 items
       0,80 €
Screw closure HDPE, red, with tamper evident ring
Product Information
  • Thread
    For
    Availability
    Price
  • 45 mm
    2, 2,5, 3, 6 and 8 L jerricans
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 0,70 €
    0,70 €
    1 * 1 items
       0,70 €
  • 50 mm
    5 and 10 L jerricans
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 0,80 €
    0,80 €
    1 * 1 items
       0,80 €
  • 60 mm
    20, 25 and 30 L jerricans
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 1,10 €
    1,10 €
    1 * 1 items
       1,10 €
Screw closure HDPE, white, with dispensing pump
Product Information
  • Thread
    For
    Availability
    Price
  • 50 mm
    5 and 10 L jerricans
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 16,30 €
    16,30 €
    1 * 1 items
       16,30 €
  • 60 mm
    20 L jerricans
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 80,40 €
    80,40 €
    1 * 1 items
       80,40 €
  • 60 mm
    25 L jerricans
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 80,40 €
    80,40 €
    1 * 1 items
       80,40 €
  • 60 mm
    30 L jerricans
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 80,40 €
    80,40 €
    1 * 1 items
       80,40 €