Barbed bulkhead fittings, Nalgene®
Supplier: Thermo Fisher Scientific
Certificates
About this item
Most Nalgene® closures and those of other manufacturers can be retrofitted with these connectors for liquid transfer without a ventilation and venting device. To do this, simply drill two fitting holes in the closure, feed in the insert and tube connectors and attach tubes with 6 or 13 mm internal diameter. Nalgene® closures 53B (53 mm) can only be fitted with the 6 mm tube connectors, the 83B (83 mm) closures can be equipped with both 6 and 13 mm connectors.
- Complete conversion kit with template and full instructions
- Leakproof assembly
- Autoclavable
PP connectors, acetal nuts, silicone seals and TPE cap.
Specifications
- Catalog No:
- 216-8931
- 216-8932
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement