VWR®, Blotting Pads, Grade 707
Supplier: VWR Collection
Certificates
About this item
This super thick paper is used to complete the blotting sandwich. Smooth, uniform texture ensures continuous contact of buffer from the chamber through the gel and transfer membrane.
- Made of 100% cotton fibre
- Produced using ultra-pure water
- Contains no additives
- Cleaner and more consistent than paper towels
Specifications
- Catalog No:
- 732-0608
- 732-0606
- 732-0604
- 732-0601
- 732-0602
- 732-0594
- 732-0599
- 732-0595
- 732-0600
- 732-0592
- 732-0597
- 732-0598
- Grade:
- 707
- 707
- 707
- 707
- 707
- 707
- 707
- 707
- 707
- 707
- 707
- 707
- Weight:
- 703 g/m²
- 703 g/m²
- 703 g/m²
- 703 g/m²
- 703 g/m²
- 703 g/m²
- 703 g/m²
- 703 g/m²
- 703 g/m²
- 703 g/m²
- 703 g/m²
- 703 g/m²
- Thickness:
- 2,60 mm
- 2,60 mm
- 2,60 mm
- 2,60 mm
- 2,60 mm
- 2,60 mm
- 2,60 mm
- 2,60 mm
- 2,60 mm
- 2,60 mm
- 2,60 mm
- 2,60 mm
- Herzberg flow rate:
- 120 s per 100 ml at 10 cm head of water
- 120 s per 100 ml at 10 cm head of water
- 120 s per 100 ml at 10 cm head of water
- 120 s per 100 ml at 10 cm head of water
- 120 s per 100 ml at 10 cm head of water
- 120 s per 100 ml at 10 cm head of water
- 120 s per 100 ml at 10 cm head of water
- 120 s per 100 ml at 10 cm head of water
- 120 s per 100 ml at 10 cm head of water
- 120 s per 100 ml at 10 cm head of water
- 120 s per 100 ml at 10 cm head of water
- 120 s per 100 ml at 10 cm head of water
- Klemm wicking rate test:
- 96 s per 7,5 cm rise
- 96 s per 7,5 cm rise
- 96 s per 7,5 cm rise
- 96 s per 7,5 cm rise
- 96 s per 7,5 cm rise
- 96 s per 7,5 cm rise
- 96 s per 7,5 cm rise
- 96 s per 7,5 cm rise
- 96 s per 7,5 cm rise
- 96 s per 7,5 cm rise
- 96 s per 7,5 cm rise
- 96 s per 7,5 cm rise