Orders LinkContactUsLinkComponent
Holder
Holder for refuse bags, white
Catalog # 129-9825
Supplier:   HUPFER METALLWERKE
Holder
Catalog # 129-9825
Supplier:  HUPFER METALLWERKE
Holder
Holder
Holder for refuse bags, white
Price
60,40 €
Log in for availability
1 * 1 items
Product Details & Documents
White steel wire with epoxy coating and rubber feet.
Int. Ø×H: 120×250 mm
Specifications
  • Description:
    Holder for refuse bags, white