Orders LinkContactUsLinkComponent
Measuring cylinders, conical, PP
Measuring cylinders, conical, PP
Supplier:   KARTELL
Measuring cylinders, conical, PP
Supplier:  KARTELL
Certificates
Measuring cylinders, conical, PP
Measuring cylinders, conical, PP
4 Options Available Below
Product Details & Documents
PP, transparent.
  • Good chemical resistance
  • Autoclavable to +121 °C
  • Liquid volume is easy to read
Ideal for a wide range of applications. With moulded graduations. Broad base for stability.
Product Family Options
Product Information
  • Capacity
    Division
    Height
    Availability
    Price
  • 100 ml
    2 ml
    118 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 6,00 €
    6,00 €
    1 * 1 items
       6,00 €
  • 250 ml
    5 ml
    164 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 9,00 €
    9,00 €
    1 * 1 items
       9,00 €
  • 500 ml
    10 ml
    183 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 10,90 €
    10,90 €
    1 * 1 items
       10,90 €
  • 1000 ml
    20 ml
    263 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 22,40 €
    22,40 €
    1 * 1 items
       22,40 €