ContactUsLinkComponent
Measuring beakers with handle, Azlon®
Measuring beakers with handle, Azlon®
Catalog # 213-0150
Supplier:  SciLabware
Supplier #: JPB3000P
undefined
Measuring beakers with handle, Azlon®
Catalog # 213-0150
Supplier:  SciLabware
Supplier #: JPB3000P
Log in for availability
Select an Option
152 mm
Select an Option
500 ml
Select an Option
3000 ml
Select an Option
240 mm
Select an Option
07
Select an Option
1
About this item
PP, highly translucent with blue graduations.
  • Robust jugs with crisp blue graduations
  • Autoclavable at +121 °C, durable with good chemical resistance
  • Ergonomically designed handle for easy carrying
Frequently Bought Together