Orders LinkContactUsLinkComponent
Beakers, low form, Nalgene®
Beakers, low form, Nalgene®
   Sustainable Options Available
Beakers, low form, Nalgene®
   Sustainable Options Available
Certificates
Beakers, low form, Nalgene®
5 Options Available Below
Product Details & Documents
PTFE/PFA, transparent, without spout.
  • Excellent chemical resistance
  • High thermal resistance from –270 to +250 °C
  • Autoclavable
Ideal for demanding applications. Not for use on hotplates. With engraved graduation. No drip, single spout design provides precision pouring control.
Specifications for Product Family
Specifications
  • Catalog No:
  • 213-0212
  • 213-7212
  • 213-7214
  • 213-7216
  • 213-7217
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
Specifications
Product Family Options
Product Information
  • Capacity
    Availability
    Price
  • 50 ml
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 56,80 €
    56,80 €
    1 * 1 items
       56,80 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    213-0212
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 100 ml
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 66,10 €
    66,10 €
    1 * 1 items
       66,10 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    213-7212
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 250 ml
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 153,00 €
    153,00 €
    1 * 1 items
       153,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    213-7214
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 600 ml
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 218,00 €
    218,00 €
    1 * 1 items
       218,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    213-7216
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 1000 ml
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 322,00 €
    322,00 €
    1 * 1 items
       322,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    213-7217
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact