Orders LinkContactUsLinkComponent
Adapters, receiver, Quickfit®
19/26 NS, 180 mm
Catalog # 201-0620
Supplier:   SciLabware
Adapters, receiver, Quickfit®
Catalog # 201-0620
Supplier:  SciLabware
Adapters, receiver, Quickfit®
Adapters, receiver, Quickfit®
19/26 NS, 180 mm
Price
27,40 €
Log in for availability
1 * 1 items
Product Details & Documents
PYREX®, borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Straight delivery.
Specifications
  • Socket size:
    19/26 NS
  • Total length:
    180 mm