Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Environmental Test Kits
Naval Fluid Colour Guide
Naval Fluid Colour Guide
Naval Fluid Colour Guide
Catalog # MILFXX6504713
Supplier:
Merck
CAS Number:
Expand image
Naval Fluid Colour Guide
Catalog # MILFXX6504713
Supplier:
Merck
CAS Number:
Specifications
Pk:
1
Description:
Naval fluid colour guide
Specifications
About this item
Product Details
Product Details