About this item
Millicup™-FLEX disposable filtration kit combines the convenience of disposable filtration with the flexibility and compatibility of traditional vacuum filtration glassware.
- Compatible with organic and aqueous solvents
- Ergonomic, clampless design
- Reduce contamination risk
- Filter directly into vacuum-rated storage bottles
- Easy access to membrane after filtration
- Fully recyclable components
- Color: Clear with blue adapter
With solvent-resistant, modular components, the Millicup-FLEX filtration unit offers a safe and efficient means for filtering a variety of aqueous and organic solutions, allowing the user to filter directly into GL 45 vacuum-rated filtration bottles with their choice of 47 mm disc membranes. The ready-to-assemble, clampless design of the Millicup-FLEX filtration system eliminates the need for cleaning prior to filtration - saving time and ultimately reducing the risk of sample contamination. Following filtration, the 250 ml funnel and membrane support collar easily disconnect to enable direct access to the membrane.
Available in 2 versions:
Starter kit: 10 funnels with 2 semi-reusable membrane support collars and vacuum tube adapters (MILFMCFLX4702)
Single-use kit: 10 funnels with 10 single-use membrane support collars and vacuum tube adapters (MILFMCFLX4710)
Specifications
- Catalog No:
- MILFMCFLX4710
- MILFMCFLX4702
- Type:
- Bottle top filter
- Bottle top filter
- Sample volume:
- 250 ml
- 250 ml
- Autoclavable:
- Not autoclavable
- Not autoclavable
- Membrane:
- Cut disc filters, Ø 47 mm
- Cut disc filters, Ø 47 mm