ContactUsLinkComponent
Thermal Printer for CryoMed™ and CryoPlus™ Freezers
Catalog # THBR4000665
New Product
Thermal Printer for CryoMed™ and CryoPlus™ Freezers
Catalog # THBR4000665
New Product
Specifications
  • Description:
    Thermal printer with cable assembly and RS-232 connector kit, 220 V, 50 Hz
  • Pk:
    1
Specifications
About this item
Enhance the functionality for Thermo Scientific™ CryoMed™ controlled-rate freezers with Thermo Scientific™ Thermal Printers for CryoMed and CryoPlus™ Freezers. These printers include cable assembly and RS-232 connector kit.