ContactUsLinkComponent
Tychem® 6000 F PA30L0 Apron
Tychem® 6000 F PA30L0 Apron
Catalog # DUPOTFPA30TGY00
Supplier:  DuPont
New Product
Tychem® 6000 F PA30L0 Apron
Catalog # DUPOTFPA30TGY00
Supplier:  DuPont
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Colour:
    Grey
  • Description:
    Apron model PA30L0, shin-length, neck and waist ties, no seams
  • Pk:
    25
Specifications
About this item
DuPont™ Tychem® 6000 F Apron model PA30L0. Shin-length apron with neck and waist ties. Available in grey and in one size.
Specially designed for use with Tychem® apparel, Tychem® accessories can help offer enhanced protection for body parts that are more exposed to hazardous substances.
Tychem® 6000 F garments and accessories are made from a lightweight and durable fabric consisting of a proprietary barrier film laminated to a heavy duty Tyvek® substrate. They offer excellent chemical permeation protection to an extensive range of chemicals, helping to protect personnel against numerous toxic industrial organic chemicals, highly concentrated inorganic chemicals (even under pressure), particulates, biohazards and certain chemical warfare agents.
Tychem® 6000 F is used for a broad range of applications from chemical spill clean-up, emergency response, to military and petrochemical applications.
Certifications: Certified according to Regulation (EU) 2016/425.Partial body chemical protective clothing, Category III, Type PB [3-B]. EN 14126 (barrier to infective agents).