Orders LinkContactUsLinkComponent
Orange Serum Agar
Orange Serum Agar
Orange serum agar, 25×200 ml bottles, 25
Catalog # LIOF452690
Supplier:   LIOFILCHEM
Orange Serum Agar
Catalog # LIOF452690
Supplier:  LIOFILCHEM
CAS Number:  
Orange Serum Agar
Orange serum agar, 25×200 ml bottles, 25
CAS Number:  
Price
237,00 €
Log in for availability
1 * 25 items
Product Details & Documents
Orange Serum Agar is a medium used for the isolation, cultivation and enumeration of microorganisms associated with spoilage of citrus fruits and derivates, such as lactic acid and acetic acid bacteria, yeasts and moulds.
Specifications
Specifications
  • Pk:
    25
  • Description:
    Orange serum agar, 25×200 ml bottles