Bottles, narrow neck, round, LDPE, with screw cap
Catalog # 215-0928
Supplier: Kautex
Specifications
- Ext.Ø:97,5 mm
- Thread:28 mm
- Capacity:1000 ml
- Height:223,5 mm
- Neck I-Ø:21,5 mm
- Pk:1
Specifications
About this item
LDPE, natural colour.
- Leak-proof, tough and versatile
- Squeezable, so these bottles make excellent containers for dripping or squirting
- Good reset properties
Excellent for sample collection, storing and transporting liquids.