ContactUsLinkComponent
Conductivity standard 147 µS/cm (25 °C; 0,001 mol/L; KCl) for conductivity measurement
Conductivity standard 147 µS/cm (25 °C; 0,001 mol/L; KCl) for conductivity measurement
  SCOI285126503
 :  
Conductivity standard 147 µS/cm (25 °C; 0,001 mol/L; KCl) for conductivity measurement
  SCOI285126503
 :  
  • Pack type:
    Glass ampoule
  • Pk:
    18 Ampoul
  • Reference temperature:
    25 °C
  • Description:
    Conductivity standard 147 µS/cm (0,001 mol/l; KCl)