ContactUsLinkComponent
Conductivity standard 1,413 µS/cm (25°C; 0,01 mol/L; KCl) for conductivity measurement
Conductivity standard 1,413 µS/cm (25°C; 0,01 mol/L; KCl) for conductivity measurement
Catalog # SCOI285136907
Supplier:  SI Analytics
CAS Number:  
Conductivity standard 1,413 µS/cm (25°C; 0,01 mol/L; KCl) for conductivity measurement
Catalog # SCOI285136907
Supplier:  SI Analytics
CAS Number:  
Specifications
  • Pack type:
    Glass bottle
  • Pk:
    250 mL
  • Reference temperature:
    25 °C
  • Description:
    Conductivity standard 1413 µS/cm (0,01 mol/l; KCl)