ContactUsLinkComponent
Conductivity standard 10,000 µS/cm (25°C; 0,0765 mol/l; KCl) for conductivity measurement
  YSIA060911
 :  
Conductivity standard 10,000 µS/cm (25°C; 0,0765 mol/l; KCl) for conductivity measurement
  YSIA060911
 :  
  • Pack type:
    Glass bottle
  • Pk:
    473 mL
  • Reference temperature:
    25 °C
  • Description:
    Conductivity standard 10 000 µS/cm (0,0765 mol/l; KCl)