ContactUsLinkComponent
Standard for ViscoQC Viscometers
Standard for ViscoQC Viscometers
Catalog # PAAR191118
Supplier:  Anton Paar
New Product
Standard for ViscoQC Viscometers
Catalog # PAAR191118
Supplier:  Anton Paar
New Product
Specifications
  • Description:
    Silicone viscosity standard AP RT5k, 5,6/5,0 Pa.s at 20/25 °C
  • Pk:
    600 mL
Specifications
About this item
Silicone viscosity standard for use with ViscoQC.
Compatible with: ViscoQC 100 H, ViscoQC 100 L, ViscoQC 100 R, ViscoQC 300 H, ViscoQC 300 L, ViscoQC 300 R