ContactUsLinkComponent
PHAGO'GEL Disinfectant Gel in Airless Bottle Pump
PHAGO'GEL Disinfectant Gel in Airless Bottle Pump
Catalog # PHAGSA0135720
Supplier:  PHAGOGENE
New Product
PHAGO'GEL Disinfectant Gel in Airless Bottle Pump
Catalog # PHAGSA0135720
Supplier:  PHAGOGENE
New Product
Specifications
  • Capacity:
    1000 ml
  • Description:
    PHAGO'GEL SPS Disinfectant gel with Airless Bottle Pump, Clear
  • Sterility:
    Sterile
  • Pk:
    12
Specifications
About this item
Intended for the hygienic surgical disinfection of the hands by friction.
  • High tolerance
  • Speed ​​of use
  • Bactericidal, fungicidal, mycobactericidal and virucidal
  • Fragrance-free, dye-free, phenoxyethanol-free, and paraben-free
It is used as an alternative to simple or hygienic hand washing, in the absence of a water point, during serial contact, before any aseptic gesture.