Large Nitrogen Dewar Flask for Bruker
Suprasil offset nitrogen dewar flask for bruker, 150 ml
Catalog # WILMWG-853-B-Q
Supplier: SP Industries
Jump to:
Product Details & Documents
These dewar Flasks provide a convenient alternative to VT systems for studies at 77 K, and are silvered around the reservoir. 150 ml flask lasts 30 minutes after a filling. The offset versions (WG-853) are designed for cavities that have limited space between the reservoir and the drive shaft for iris.
Specifications
- Description:Suprasil offset nitrogen dewar flask for bruker, 150 ml