EasySep™ Human Th17 Cell Enrichment Kit II
Catalog # STMC17862
Supplier: STEMCELL Technologies
Specifications
- Description:EasySep™ human Th17 cell enrichment kit II, 2×10⁹ cells
- Environmentally Preferable:
- Pk:1 KIT
Specifications
About this item
Immunomagnetic positive selection of human Th17 cells.
- Fast and easy-to-use
- Up to 98% purity
- No columns required
The EasySep™ Human Th17 Enrichment Kit II is a two-step method designed to enrich for human Th17 cells from fresh peripheral blood mononuclear cells. First, CCR6+ cells are isolated by column-free immunomagnetic positive selection using antibody complexes and EasySep™ Releasable RapidSpheres™. Then, bound magnetic particles are removed from the cells and unwanted non-CD4+ T cells, CD45RA+ cells, and CXCR3+ cells are targeted for depletion using antibody complexes and EasySep™ Dextran RapidSpheres™. The final isolated fraction is enriched for IL-17-producing Th17 cells.
This product can be used in place of the EasySep™ Human Th17 Cell Enrichment Kit for even faster cell isolations.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').