ContactUsLinkComponent
EasySep™ Mouse/Human Chimera Isolation Kit
EasySep™ Mouse/Human Chimera Isolation Kit
  STMC19849
undefined
EasySep™ Mouse/Human Chimera Isolation Kit
  STMC19849
  • Description:
    EasySep™ mouse/human chimera isolation kit, 1×10⁹ cells
  • Pk:
    1 KIT
Immunomagnetic isolation of untouched human cells
  • Fast, easy-to-use and column-free
  • Isolated cells are untouched
The EasySep™ Mouse/Human Chimera Isolation Kit is designed to highly enrich human cells from bone marrow, spleen or peripheral blood of human xenograft recipient mice by negative selection. Unwanted cells are targeted for removal with biotinylated antibodies directed against mouse hematopoietic cells and streptavidin-coated magnetic particles. labelled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.
This product can be used in place of the StemSep™ Mouse/Human Chimera Enrichment Kit (Catalog #13068) for faster, easier cell isolations.
 : Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').