ContactUsLinkComponent
EasySep™ Human Bone Marrow CD138 Positive Selection Kit
EasySep™ Human Bone Marrow CD138 Positive Selection Kit
Catalog # STMC100-1133
undefined
EasySep™ Human Bone Marrow CD138 Positive Selection Kit
Catalog # STMC100-1133
Specifications
  • Description:
    EasySep™ human bone marrow CD138 positive selection kit, 1 unit
  • Pk:
    1 KIT
Specifications
About this item
Immunomagnetic positive selection of human CD138+ cells from bone marrow samples for in vitro diagnostic (IVD) applications.
  • Isolate plasma cells quickly and reliably
  • Follow an easy-to-use and column-free protocol
  • Reduce your validation burden by using an IVD product
  • Enhance the sensitivity of downstream fluorescence in situ hybridisation (FISH)
Reduce the validation burden for your laboratory and effectively and reliably enrich plasma cells with the EasySep™ Human Bone Marrow CD138 Positive Selection Kit. This kit is an in vitro diagnostic and EasySep™ Buffer to isolate cells without the use of columns, the EasySep™ Human Bone Marrow CD138 Positive Selection Kit includes the antibodies and magnetic particles required to label CD138+ cells for separation. The easy-to-follow protocol results in labeled CD138+ cells that remain in the tube after unlabeled cells are poured off. This kit is indicated for in vitro diagnostic use by laboratory professionals; the end user is responsible for determining whether the product is suitable for their specific application.
Note: Ref #100-0748 is the Regulatory Product ID and should not be used for ordering.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the ‘Product Restrictions’).