Order Entry
Slovenia
ContactUsLinkComponent
NMPH Solution (1 ml (1 mg/ml)) (Methyl 2-(1-nitrosopiperidin-2-yl)-2-phenylacetate), Pharmaceutical Analytical Impurities
NMPH Solution (1 ml (1 mg/ml)) (Methyl 2-(1-nitrosopiperidin-2-yl)-2-phenylacetate), Pharmaceutical Analytical Impurities
Catalog # 1A03940
Supplier:  USP
New Product
CAS Number:  
NMPH Solution (1 ml (1 mg/ml)) (Methyl 2-(1-nitrosopiperidin-2-yl)-2-phenylacetate), Pharmaceutical Analytical Impurities
Catalog # 1A03940
Supplier:  USP
New Product
CAS Number:  

Specifications

  • Pk:
    1
  • Description:
    N-Nitrosomethylphenidate (NMPH) solution (1 ml (1 mg/ml)) (Methyl 2-(1-nitrosopiperidin-2-yl)-2-phenylacetate)
  • UN No.:
    1230
  • GHS signal:
    Danger
  • Formula:
    C₁₄H₁₈N₂O₃

Specifications

About this item