Copper tetramine complex in aqueous solution (copper(II) hydroxide 10% in ammonia solution 20%) Reag. Ph. Eur. 1022600
Catalog # 87809.180
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 mL
- Seq:2440
Specifications
About this item
:
:97.56 g/mol
EINECS:243-815-9
UN:3266
ADR:8,II
:GHS05
:GHS09
CAS Number:20427-59-2
Formula:Cu(OH)₂