FitQuik™ Tubing Connectors, Straight, Reducer
Catalog # CPCCHSR128
Supplier: CPC COLDER
New Product
Specifications
- Description:Polypropylene reducer fitting ³/₈ to ¹/₄" hose barb
- Pk:100
Specifications
About this item
FitQuik™ Connectors from Colder Products Company are high quality fittings for leak-free tubing connections. These precision molded fittings are designed to eliminate tubing leak points in applications such as medical devices, analytical instrumentation or air-driven equipment.
Cost-competitive FitQuik connectors are offered in many standard configurations or in custom designs to meet unique customer specifications.
Quality FitQuik™ connectors are instrumentation grade fittings that ensure a secure fit in even the most demanding applications. These fittings feature precise hose barbs and thread forms to ensure the right fit, every time.