ContactUsLinkComponent
CHROMABOND® WAX SPE Columns
CHROMABOND® WAX SPE Columns
Supplier:  MACHEREY-NAGEL
New Product
Certificates
CHROMABOND® WAX SPE Columns
Supplier:  MACHEREY-NAGEL
New Product
Specifications
  • Catalog No:
  • MANA7300015
  • MANA7300014
  • pH range:
  • 1 - 14
  • 1 - 14
  • Surface area:
  • 800 m²/g
  • 800 m²/g