ContactUsLinkComponent
Cif Pro Formula SafeGuard Combined Cleaner Disinfectants
Cif Pro Formula SafeGuard Combined Cleaner Disinfectants
Catalog # DIVY101108148
Supplier:  Diversey
New Product
Cif Pro Formula SafeGuard Combined Cleaner Disinfectants
Catalog # DIVY101108148
Supplier:  Diversey
New Product
Specifications
  • Capacity:
    6×0,75 L
  • Description:
    Cif Pro Formula Safeguard disinfectant
  • Seq:
    02
  • Pack type:
    Bottle
  • Pk:
    6 Bottle
Specifications
About this item
Cif Pro Formula SafeGuard 2in1 cleaner disinfectant is a highly concentrated detergent disinfectant for one step cleaning and disinfection of all hard surfaces including in food premises. It is very effective against a wide range of micro-organisms, in all water conditions and cuts through grease and dried-on food soiling.
  • Kills bacteria and enveloped viruses (including Coronaviruses)
  • Powerful cleaner and disinfectant
  • Cuts through grease and dried-on food soiling
  • Suitable for use on hard surfaces inside and outside the kitchen
  • Perfume free
Perfume free disinfection ideal for food premises. Effective agains a wide range of micro-organisms. Cuts through grease and dried on food soiling easily.
Certifications: BS EN 1276 Accredited with 30 seconds contact time; EN14476 with one minute.