Thread Adapter, GL 32
Thread adapter, Borosilicate glass 3.3, Transparent
Catalog # NEOLE-2933
Supplier: NEOLAB
Jump to:
Product Details & Documents
These are required to attach threaded vials to ground joint apparatus such as rotary evaporators.
Ordering information: Size-dependent screw couplings are required for this purpose.
Specifications
- Colour:Transparent
- Description:Thread adapter
- Material:Borosilicate glass 3.3