HIS Lite™ Cy3 Bis NTA Chelator
Catalog # AATB12650
Supplier: AAT BIOQUEST
New Product
CAS Number:
Specifications
- Pk:100 µG
- Description:HIS Lite™ Cy3 Bis NTA chelator
Specifications
About this item
Cy3 Bis NTA-Ni Chelator can be applied either to an SDS-PAGE gel or Western blot membrane for fluorescence imaging or used in living cells in combination with the addition of a heavy metal ion such as Ni²⁺.
- Stable for 24 months
Quality tested in-house.
Caution: For research use only.