Buccutite™ Rapid Oligo Antibody Conjugation Kit
Catalog # AATB5450
Supplier: AAT BIOQUEST
New Product
CAS Number:
Specifications
- Pk:2
- Includes:2 labellings
Specifications
About this item
Optimised for labelling 100 µg protein. Buccutite™ rapid oligo antibody conjugation kit is optimised for conjugating 100 µg protein with an amino-modified oligo.
- Robust protocol
- Consistent results
- 12-month shelf life
Designed to provide high quality material for downstream fluorescent applications, including flow cytometry.
Delivery: Shipped at RT.
Caution: Research use only.