Orders LinkContactUsLinkComponent
HIS Lite™ Cy5 Tris NTA-Ni Complex
HIS Lite™ Cy5 Tris NTA-Ni Complex
HIS Lite™ Cy5 tris NTA-Ni complex, 100 µG
Catalog # AATB12619
Supplier:   AAT Bioquest
HIS Lite™ Cy5 Tris NTA-Ni Complex
Catalog # AATB12619
Supplier:  AAT Bioquest
CAS Number:  
HIS Lite™ Cy5 Tris NTA-Ni Complex
HIS Lite™ Cy5 Tris NTA-Ni Complex
HIS Lite™ Cy5 tris NTA-Ni complex, 100 µG
CAS Number:  
Price
275,00 €
Log in for availability
1 * 100 µG
Product Details & Documents
Cy5-Tris NTA compound is used as a sensitive fluorescent probe for detecting polyhistidine-labelled proteins in cells, solution and solid surfaces.
  • Stable for 12 months
Quality tested in-house.
Caution: For research use only.
Specifications
Specifications
  • Pk:
    100 µG
  • Description:
    HIS Lite™ Cy5 tris NTA-Ni complex