TAQuest™ qPCR Master Mix for TaqMan Probes *Low ROX*
Supplier: AAT Bioquest
New Product
Certificates
About this item
TAQuest™ qPCR Master mix for TaqMan probes is a ready-to-use 2X solution optimised for qPCR and two-step RT-qPCR compatible with TaqMan gene expression assays.
- Simple and robust protocol
- The optimised composition ensures PCR specificity and sensitivity with all sample types such as genomic, plasmid, viral and cDNA templates
TAQuest™ qPCR Master mix for TaqMan probes has been designed to be used for duplex reactions using internal positive controls.
Caution: Master mix is stable for 12 months when stored frozen (<−15 °C) with minimised light exposure.
Specifications
- Application:qPCR probe